Learn More
Abcam Bezafibrate, Pan-PPAR Agonist, Solid, >99% Purity, 5 g
Description
- Molecular Weight: 361.8 Da
- Purity: >99%
- Form: Solid
- Mechanism: Pan-PPAR agonist (EC50 values are 50, 60 and 20 μM at PPARα, PPARγ and PPARδ human receptors respectively)
- Biological Effects: Antidiabetic and antihyperlipidemic effects
- Storage: Store at ambient temperature
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | PPARα, PPARγ, PPARδ |
| Shipping Condition | Ambient |
| Buffer | 100% Bezafibrate |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Formulation | 100% Bezafibrate |
| Quantity | 5 g |
| For Use With (Application) | Antidiabetic research, Antihyperlipidemic research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.