Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BFAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596998 Supplier Invitrogen™ Supplier No. PA596998
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEPQKSYVN TMDLERDEPL KSTGPQISVS EFSCHCCYDI LVNPTTLNCG HSFCRHCLAL WWASSKKTEC PECREKWEGF PKVSILLRDA IEKLFPDAIR LRFEDIQQNN DIVQSLAAFQ KYGNDQIPLA PNTGRANQQM; Positive Samples: HepG2, SKOV3, A-549, SH-SY5Y, SW480, Mouse brain, Mouse heart, Mouse liver, Rat heart; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.

The bifunctional apoptosis inhibitor (BFAR) is scaffold protein that integrates signaling components of the cells apoptosis-regulatory machinery. BFAR is a multidomain protein capable of inhibiting apoptosis induced by TNF-family death receptors ('extrinsic pathway') as well as mitochondria-dependent apoptosis ('intrinsic pathway'). Interaction of BFAR with Bcl-2 or Bcl-XL via a SAM domain may contribute to the anti-apoptotic properties of BFAR. In addition, BFAR contains a DED-like domain that is capable of suppressing apoptosis mediated at the receptor level. BFAR is also thought to be involved in the regulation of neuronal survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BFAR
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BFAR
Gene Accession No. Q5PQN2, Q8R079, Q9NZS9
Gene Alias 3010001A07Rik; AI666707; AW107665; BAR; Bfar; bifunctional apoptosis inhibitor; bifunctional apoptosis regulator; RING finger protein 47; RNF47
Gene Symbols BFAR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human BFAR (NP_0576451).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304709, 51283, 67118
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.