Learn More
Cayman Chemical BGJ398phosphate 25mg
SDP
Supplier:  Cayman Chemical 3929225 MG
Encompass_Preferred
A prenylated coumarin with anti-inflammatory and neuroprotective activities inhibits TPA- and A23187-induced histamine release from HMC-1 cells at 10 UM protects against glutamate-induced cytotoxicity in primary rat cortical cells at 0.1-10 UM
Catalog No. 50-291-7868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.