Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BGP 15
GREENER_CHOICE

Catalog No. p-7166819
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-035 5 mg
67-032-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-035 Supplier Tocris Bioscience™ Supplier No. 6703/5
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

PARP inhibitor; cytoprotectant

BGP 15 is a PARP inhibitor (IC50 = 120 μM). Displays cytoprotective effects in various disease models: decreases ROS levels, lipid peroxidation and single-strand DNA breaks in isolated reperfused rat hearts; improves mitochondrial function and protects against neuronal cell death in a mouse model of familial dysautonomia. Also activates HSP70 in rodent skeletal muscle, increasing oxidative capacity and improving insulin sensitivity.

Specifications

Quantity 5 mg
Molecular Formula C14H22N4O2.2HCl
Formula Weight 351.27
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.