Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHA, FCC, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18600970 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-970 500 g Amber Glass Bottle
18-600-969 125 g Amber Glass Bottle
18-600-971 2.5 kg Amber Glass Bottle
18-600-972 12 kg Amber Glass Bottle
18-600-973 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-970 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BH104500GM
Only null left
Add to Cart
Add to Cart

BH104, 25013-16-5, C11H16O2

Spectrum™ Chemical BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES COC1=CC=C(O)C(=C1)C(C)(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98.5%
Grade FCC
Physical Form Crystals or Flakes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.