Learn More
BioMed Diagnostics Offers the fastest read time for BHB in bovine blood. Five seconds for BHB results, seven seconds for glucose. | Detects: Blood ketone and glucose in bovine. | Available in: 6 case
Supplier:  BioMed Diagnostics HM006
Encompass_Preferred
The award-winning BHBCheck Plus meter offers the fastest read time for BHB in bovine blood. This amazing little meter can read BHB and glucose in blood. Five seconds for BHB results, seven seconds for glucose. The University of Wisconsin-Madison validated our meter, and they use it for all of their herd work. | Detects: Blood ketone and glucose in bovine. | Available in: Case of 6
Catalog No. NC2582822
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.