Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam BHHCT, Fluorescent Sensitizer, >90% Purity, Powder, 1 g
Description
- Molecular Weight: 804.9 Da
- Purity: >90%
- Form: Powder
- Application: Fluorescent sensitizer for Eu3+ labeling; measures peroxidase activity and oxidative stress levels
- Performance Characteristics: Increases luminescence intensity 11-fold; persists as an Eu3+ complex for 500 μs
- Composition: 100% BHHCT
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Eu3+ |
| Substrate Properties | Chemical Substrate |
| Substrate | Peroxidase |
| Shipping Condition | Ambient |
| Label or Dye | Eu3+ Labeling Sensitizer |
| Form | Powder |
| Buffer | 100% BHHCT |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Formulation | 100% BHHCT |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?