Learn More
Abcam BHHCT, Fluorescent Sensitizer, >90% Purity, Powder, 100 mg
Description
- Molecular Weight: 804.9 Da
- Purity: >90%
- Physical Form: Powder
- Function: Fluorescent sensitizer for Eu3+ labelling; increases luminescence intensity 11-fold
- Applications: Measures peroxidase activity and oxidative stress levels
- Stability: Persists as an Eu3+ complex for 500 μs
- Composition: 100% BHHCT
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Eu3+ |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Form | Powder |
| Detection Method | Fluorescence |
| Buffer | 100% BHHCT |
| Content And Storage | Store at -20°C. |
| Formulation | 100% BHHCT |
| Quantity | 100 mg |
| For Use With (Application) | Eu3+ labelling, Peroxidase activity measurement, Oxidative stress detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.