Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam BHHCT, Fluorescent Sensitizer, Eu3+ Labelling, Powder, 5 mg

Catalog No. 502705295
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5295 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5295 Supplier Abcam Supplier No. AB1453145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Increases luminescence intensity 11-fold and measures peroxidase activity and oxidative stress levels.

  • Fluorescent sensitizer for Eu3+ labelling
  • Increases luminescence intensity 11-fold
  • Measures peroxidase activity and oxidative stress levels
  • Persists as an Eu3+ complex for 500 μs
  • Molecular weight: 804.9 Da
  • Purity: >90%
  • Form: Powder

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Eu3+
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Substrate Peroxidase
Reactivity Peroxidase
Buffer 100% BHHCT
Content And Storage Store at -20°C.
Form Powder
Detection Method Fluorescence
Formulation 100% BHHCT
Quantity 5 mg
For Use With (Application) Eu3+ labelling, peroxidase activity measurement, oxidative stress detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.