Learn More
Medchemexpress LLC Bi-1622 | 2681392-19-6 | 99.0% | 508.53 g/mol | C26H24N10O2 | 10 MG
Supplier: Medchemexpress LLC HY15002310MG
BI-1622 is an orally active, potent, and highly selective inhibitor of the HER2 (ERBB2) kinase, developed for preclinical research. It has reported biochemical potency (IC50) of 7 nM, shows greater than 25-fold selectivity over EGFR, and has demonstrated efficacy in HER2-driven xenograft models.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.