Learn More
TARGETMOL CHEMICALS INC BI-1408 5MG
Supplier: TARGETMOL CHEMICALS INC T105325MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BI-1408 is a modulator potent of (gamma) secretase (IC50 0.04 uM for A(beta)42). purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.