Learn More
Description
- Used in competitive BRET assays to quantify RAS target engagement.
- Derived from the reversible KRAS inhibitor BI 2852.
Specifications
Specifications
| Quantity | 100 μg |
| Product Type | Fluorescent Probe |
| Content And Storage | Store at -20°C |
| Target | Other Fluorescent Probes |
| Label Type | Other Labels or Dyes |
| Product Line | Tocris |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
