Learn More
Selleck Chemical LLC BI-3812 2mg 2166387-64-8
SDP
Supplier:  Selleck Chemical LLC S87352MG
Encompass_Preferred
BI-3812 is a highly potent and efficacious BCL6 inhibitor probe compound with an IC50 less than 3 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1770117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.