Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BI 605906

Catalog No. 530010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 53-001-0 Supplier Tocris Bioscience™ Supplier No. 5300/10
This item is not returnable. View return policy

Selective IKKβ inhibitor

BI 605906 is a selective IKKβ inhibitor (IC50 = 380 nM). Displays no effect on a panel of 100 kinases, except at IGF1 (IC50 = 7.6 μM).

Specifications

Product Type Inhibitor
Target Proteins IkB Kinase
CAS 960293-88-3
Molecular Formula C17H22F2N4O3S2
Formula Weight 432.51g/mol
Content And Storage Store at -20°C
For Use With (Application) IkB Kinase Inhibitors
Quantity 10 mg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.