Learn More
Apexbio Technology LLC BI-D1870 501437-28-1 5mg
SDP
Supplier:  Apexbio Technology LLC B22275
Encompass_Preferred
BI-D1870 (CAS 501437-28-1) is a cell-permeable small molecule inhibitor that selectively targets the p90 ribosomal S6 kinase (RSK) family comprising RSK1 RSK2 RSK3 and RSK4 It functions as an ATP-competitive inhibitor exhibiting IC50 values of 31 nM 24 nM 18 nM and 15 nM against RSK1 RSK2 RSK3 and RSK4 respectively BI-D1870 demonstrates over 500-fold greater selectivity for RSK isoforms compared to other AGC kinases In cellular assays such as with Rat-2 cells it has been shown to induce phosphorylation of ERK1/ERK2 and subsequent CREB phosphorylation BI-D1870 is widely utilized for investigating RSK-mediated signal transduction and downstream cellular processes in biomedical research
Catalog No. 50-224-0139
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.