Learn More
Cytiva Biacore maintenance kit (type 3)
Supplier:  Cytiva 29229054
Encompass
Kit for conducting instrument maintenance of Biacore 8K+ and Biacore 8K SPR systems, containing BIAtest, BIAdesorb and BIAnormalizing solutions.
Catalog No. 50-152-8210