Learn More
Cytiva BIAnormalizing solution (70%), 90mL
Supplier:  Cytiva 29207950
Encompass
A glycerol solution to be used when performing normalization of the detector response in Biacore 8K+, Biacore 8K, Biacore S200, Biacore T200, and Biacore 4000 SPR systems.
Catalog No. 50-152-8209