Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bianthrone 95.0+%, TCI America™
SDP

Catalog No. B0827100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0827100MG 100 mg
B08271G 1 g
B08275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0827100MG Supplier TCI America Supplier No. B0827100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 434-85-5
Molecular Formula C28H16O2
Molecular Weight (g/mol) 384.434
MDL Number MFCD00001238
InChI Key MGRRGKWPEVFJSH-UHFFFAOYSA-N
Synonym bianthrone, 10h,10'h-9,9'-bianthracenylidene-10,10'-dione, dehydrodianthrone, 9 10h-anthracenone, 10-10-oxo-9 10h-anthracenylidene, .delta.10,10'-bianthrone, 10-10-oxo-9-anthrylidene anthracen-9-one, 10-10-oxoanthracen-9-ylidene anthracen-9-one, bisanthrone, 10-10-oxoanthracen-9 10h-ylidene anthracen-9 10h-one, 9,9'-bianthracenylidene-10,10'-dione
PubChem CID 67948
IUPAC Name 10-(10-oxoanthracen-9-ylidene)anthracen-9-one
SMILES C1=CC=C2C(=C1)C(=C3C4=CC=CC=C4C(=O)C5=CC=CC=C53)C6=CC=CC=C6C2=O

Specifications

Melting Point 300°C
Color Green-Yellow
Quantity 100 mg
Formula Weight 384.43
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bianthrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.