Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bianthronyl 97.0+%, TCI America™
SDP

Catalog No. D13891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13891G 1 g
D13895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13891G Supplier TCI America Supplier No. D13891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 434-84-4
Molecular Formula C28H18O2
Molecular Weight (g/mol) 386.45
MDL Number MFCD00019128
InChI Key ZQXZUOJNJXNUEO-UHFFFAOYSA-N
PubChem CID 97174
IUPAC Name 10-(10-oxo-9H-anthracen-9-yl)-10H-anthracen-9-one
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C3C2=O)C4C5=CC=CC=C5C(=O)C6=CC=CC=C46

Specifications

Melting Point 272°C
Color Yellow
Quantity 1 g
Synonym 10,10′C-Dihydrobianthrone, 10,10′C-Dihydrodianthrone
Formula Weight 386.45
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bianthronyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.