Learn More
National Safety Apparel™ Bib Overall 9 oz. Khaki Indura Ultra Soft, Multi
SDP
Catalog No. 17001363 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-363 5X-Large
17-001-357 Small
17-001-358 Medium
17-001-359 Large
17-001-360 2X-Large
17-001-361 3X-Large
17-001-362 4X-Large
17-001-364 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-363 Supplier National Safety Apparel™ Supplier No. C45UTUK255X32
Add to Cart
Add to Cart
Protective bib overalls provide the highest level of protection in work with electrical exposures and hazards.
  • FR fabric will not melt, drip or ignite
  • FR hook and loop fly closure
  • FR hook and loop calf closure for easy boot entry
  • Inherently flame resistant for FR Protection that will not wash off
  • Absorbs moisture
  • Quick disconnect and easy to adjust
  • Shoulder straps for quick entry
Specifications
Closure Type Buckle
Color Khaki
Material Cotton, Nylon
Seam Type Stitched
Size 5X-Large
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 25 cal/cm²
Waist Style Standard
Resistance Flame
Certifications/Compliance CAT 3, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462
Disposable Reusable