Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIBO 3304 trifluoroacetate
GREENER_CHOICESDP

Catalog No. 241210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-121-0 10 mg
24-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-121-0 Supplier Tocris Bioscience™ Supplier No. 2412/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly selective NPY Y1 receptor antagonist

BIBO 3304 trifluoroacetate is a high affinity NPY Y1 receptor antagonist (IC50 values are 0.38 and 0.72 nM at human and rat receptors respectively) that displays > 2600-fold selectivity over Y2, Y4 and Y5 receptors. Inhibits NPY - and fasting-induced feeding in vivo following central administration. Also antagonizes anxiolytic-like effects of NPY.

Specifications

Chemical Name or Material N-[(1R)-1-[[[[4-[[(Aminocarbonyl)amino]methyl]phenyl]methyl]amino]carbonyl]-4-[(aminoiminomethyl)amino]butyl]-α-phenyl-benzeneacetamide ditrifluoroacetate
CAS 2310085-85-7
Assay Percent Range >98%
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C29H35N7O3.2CF3CO2H
Formula Weight Observed MW: 757.69

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.