Learn More
Selleck Chemical LLC BIBO 3304 Trifluoroacetate
SDP
Supplier:  Selleck Chemical LLC E00145MG
Encompass_Preferred
BIBO 3304 Trifluoroacetate (BIBO-3304 TFA) is a novel neuropeptide Y Y1 receptor (NPY1R) antagonist with IC50 values of 0 38 nM and 0 72 nM for human Y1 receptor and rat Y1 receptor respectively BIBO 3304 shows low affinity for the human Y2 receptor human and rat Y4 receptor as well as for the human and rat Y5 receptor (IC50 values 1000 nM)
Catalog No. 50-234-0981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.