Learn More
Selleck Chemical LLC BIBR 1532 50mg 321674-73-1
SDP
Supplier:  Selleck Chemical LLC S118650MG
Encompass_Preferred
BIBR 1532 is a potent, selective, non-competitive telomerase inhibitor with IC50 of 100 nM in a cell-free assay. No inhibition of DNA and RNA polymerases, including HIV reverse transcriptase are observed at concentrations vastly exceeding the IC50 for telomerase. BIBR 1532 induces apoptosis in cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-887-7
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.