Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicalutamide 98.0+%, TCI America™
SDP

Catalog No. B32061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32061G 1 g
B3206200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32061G Supplier TCI America Supplier No. B32061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90357-06-5
Molecular Formula C18H14F4N2O4S
Molecular Weight (g/mol) 430.374
MDL Number MFCD00869971
InChI Key LKJPYSCBVHEWIU-UHFFFAOYSA-N
Synonym bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex
PubChem CID 2375
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
SMILES CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O

Specifications

Melting Point 196°C
Color White
Quantity 1 g
Formula Weight 430.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bicalutamide
RTECSNumber TX1413500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.