Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BICC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116342
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116342 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116342 Supplier Invitrogen™ Supplier No. PA5116342
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BICC1 Polyclonal Antibody for Western Blot

Putative RNA-binding protein. Acts as a negative regulator of Wnt signaling. May be involved in regulating gene expression during embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BICC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene BICC1
Gene Accession No. Q9H694
Gene Alias bicaudal C homolog 1; BICC; Bic-C; BicC family RNA binding protein 1; BICC1; CYSRD; FGFR2-BICC1 fusion kinase protein; protein bicaudal C homolog 1
Gene Symbols BICC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 75-107 amino acids from the N-terminal region of human BICC1.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.