Learn More
Selleck Chemical LLC Bictegravir Sodium-E0161-25MG
SDP
Supplier:  Selleck Chemical LLC E016125MG
Encompass_Preferred
Bictegravir (GS-9883) Sodium is a novel and potent inhibitor of HIV-1 integrase specifically targets IN strand transfer activity with an IC50 of 7 5 nM
Catalog No. 50-313-6855
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.