Learn More
Apexbio Technology LLC (-)-Bicuculline methiodide(Synonyms: (-)-Bicuculline methiodide, Bicuculline methiodide), 50mg, CAS: 40709-69-1.
SDP
Supplier:  Apexbio Technology LLC B708150
Encompass_Preferred
(-)-Bicuculline methiodide (CAS 40709-69-1) is an antagonist of the -aminobutyric acid-A (GABA A) receptor inhibiting GABA binding with an IC 50 of approximately 500 M GABA A receptors are ligand-gated chloride ion channels mediating inhibitory neurotransmission in the central nervous system In studies using chick cerebellar synaptic membranes this enantiomer exhibited lower activity relative to the ( )-enantiomer Additionally in C elegans (-)-bicuculline methiodide helped demonstrate pharmacological distinctions between UNC-49 GABA receptors and mammalian GABA A receptors This compound is employed widely in neuropharmacological research exploring GABA receptor function and regulation
Catalog No. 50-000-61264
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.