Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Bicycle Wheel Gyroscope
SDPSCI_ED

Catalog No. S27790 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Activity Guide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27790 Activity Guide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27790 Supplier United Scientific Supplies Supplier No. BWG001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrates gyroscopic forces, angular momentum, and torque

  • Constructed from durable, molded plastic
  • Better distortion resistance than a spoked wheel
  • Features easy-grip handles and free-running bicycle bearings to allow for longer demonstrations
  • Includes activity guide

Specifications

Product Type Gyroscope
For Use With (Application) Demonstrating Gyroscopic Forces, Angular Momentum, and Torque
Includes Activity Guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.