Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[1.1.1]pentan-1-amine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B54791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54791G 1 g
B5479200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54791G Supplier TCI America Supplier No. B54791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22287-35-0
Molecular Formula C5H10ClN
Molecular Weight (g/mol) 119.592
MDL Number MFCD28166320
InChI Key LQKLVOWNBKJRJE-UHFFFAOYSA-N
PubChem CID 54589873
IUPAC Name bicyclo[1.1.1]pentan-3-amine;hydrochloride
SMILES C1C2CC1(C2)N.Cl

Specifications

Melting Point 240°C
Color White
Quantity 1 g
Formula Weight 119.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bicyclo[1.1.1]pentan-1-amine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.