Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde 95.0+%, TCI America™
SDP

Catalog No. B52791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52791G 1 g
B5279200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52791G Supplier TCI America Supplier No. B52791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38259-00-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
InChI Key KCHFYKPIADOPBJ-UHFFFAOYSA-N
PubChem CID 12493694
IUPAC Name bicyclo[2.2.2]oct-2-ene-5-carbaldehyde
SMILES C1CC2C=CC1CC2C=O

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 73°C
Quantity 1 g
Formula Weight 136.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.