Learn More
BIG1/ARFGEF1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300998A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
The function of this protein remains unknown.Specifications
| BIG1/ARFGEF1 | |
| Polyclonal | |
| Unconjugated | |
| ARFGEF1 | |
| ADP-ribosylation factor guanine nucleotide-exchange factor 1, ADP-ribosylation factor guanine nucleotide-exchange factor 1 (brefeldin A-inhibited), ARFGEP1, BIG1, Brefeldin A-inhibited GEP 1, Brefeldin A-inhibited guanine nucleotide-exchange protein 1, DKFZP434L057, p200, p200 ARF guanine nucleotide exchange factor, p200 ARF-GEP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10565, 211673 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| G3X9K3, Q9Y6D6 | |
| Arfgef1 | |
| Between 1800 and C-term | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.