Learn More
Apexbio Technology LLC E-4031 dihydrochloride 113559-13-0 5mg
SDP
Supplier:  Apexbio Technology LLC B68625
Encompass_Preferred
E-4031 dihydrochloride (CAS 113559-13-0) is a methanesulfonanilide compound classified as a class III antiarrhythmic agent It selectively inhibits ether- -go-go-related gene (ERG) family potassium channels including hERG (Kv11 1) with an IC50 of 29 nM and demonstrates binding affinities of 99 116 and 193 nM for ERG1 ERG2 and ERG3 respectively By blocking these voltage-gated K channels E-4031 dihydrochloride extends cardiac action potential duration and modulates inwardly rectifying potassium currents in myocardial and gastrointestinal cells This molecule is widely used to study ERG channel physiology arrhythmogenesis and pharmacological rescue of mutant hERG in long QT syndrome type 2
Catalog No. 50-000-88451
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.