Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BIIE-0246 hydrochloride | 246146-55-4 | MFCD04113026 | >98.0% | 896.05 g/mol | C49H57N11O6 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10198650MG

Encompass_Preferred

BIIE-0246 is a selective, non-peptide antagonist of the neuropeptide Y Y2 receptor used as a research tool in receptor pharmacology and preclinical studies. It is typically supplied as the hydrochloride salt and is appropriate for in vitro assays and parenteral in vivo investigations.

  • Selective antagonist of the NPY Y2 receptor.
  • Potent activity (IC50 ≈ 15 nM).
  • Typically supplied as a hydrochloride salt for lab use.
  • Purity ≥98.0% by HPLC.
  • Molecular weight 896.05 g/mol.
  • Available in mg-scale quantities for research applications.
  • Intended for research use only; not for human therapeutic use.

Catalog No. 50-002-48683


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.