Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bile Esculin Azide Agar
GSA_VA

Catalog No. R01189
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
12mm x 85mm Monoplate
Media Type:
Bile Esculin Azide Agar
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type
R01189 100/Pk. 12mm x 85mm Monoplate Bile Esculin Azide Agar
R01194 10/Pk. 12mm x 85mm Monoplate Bile Esculin Azide Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01189 Supplier Thermo Scientific™ Supplier No. R01189
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Isolate, enumerate and presumptively identify group D streptococci and enterococci with a medium that utilizes bile salts to improve selectivity.

Isolate, enumerate and presumptively identify group D streptococci and enterococci using Thermo Scientific™ Bile Esculin Azide Agar, a microbiological media that includes a high portion of Ox bile.

Ox bile present in the media inhibits the growth of Gram-positive bacteria except Enterococci, while Sodium azide suppresses the Gram-negative bacteria. Group D streptococci and enterococci are able to hydrolyse aesculin into aesculetin and glucose, and the bondage of aesculetin and ferric ammonium citrate causes the brown-black to black halo around the colonies.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, SMWW.
Format 12mm x 85mm Monoplate
Manufacturing Origin United States
Media Type Bile Esculin Azide Agar
Product Type Agar Plate
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.