Learn More
Bioassay Systems QuantiChrom™ Bilirubin Assay Kit. For quantitative determination of bilirubin and evaluation of drug effects on bilirubin metabolism.
SDP
Catalog No. 50489210 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of bilirubin and evaluation of drug effects on bilirubin metabolism. Key Features: Sensitive and accurate. Detection limit 0.16 mg/dL bilirubin in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 10 min. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Method: OD530nm. Samples: Serum. Species: All. Procedure: Assay takes 10 min. Kit size: 180 tests. Detection limit: 0.16 mg/dL.
Catalog No. 50-489-210 Supplier Bioassay Systems Supplier No. DIBR180
Add to Cart
Add to Cart
For quantitative determination of bilirubin and evaluation of drug effects on bilirubin metabolism. Key Features: Sensitive and accurate. Detection limit 0.16 mg/dL bilirubin in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 10 min. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Method: OD530nm. Samples: Serum. Species: All. Procedure: Assay takes 10 min. Kit size: 180 tests. Detection limit: 0.16 mg/dL.