Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bilirubin Oxidase, MP Biomedicals™

Catalog No. MP219000020 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219000020 20 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219000020 Supplier MP Biomedicals™ Supplier No. 0219000020
Add to Cart
Add to Cart

Catalyzes the oxidation of bilirubin to biliverdin. Bilirubin Oxidase, MP Biomedicals is a multicopper oxidase enzyme that is used for diagnostic analysis of bilirubin in serum during medical examinations of the serum. Oxygen is the electron acceptor and

Bilirubin oxidase is a monomeric protein with a molecular mass of 60 kDa and high reactivity at neutral pH. BOD participates in porphyrin and chlorophyll metabolism. It is used to degrade bilirubin and for numerous laboratory applications.

  • Bilirubin oxidase, from Myrothecium verrucaria, may be used to determine free hemoglobin in icteric specimens
  • It has potential application in dye effluent decolorization
  • It is a potential treatment for neonatal jaundice
  • It is used diagnostically to determine two types of bilirubin, glucuronide-conjugated and unconjugated bilirubin separately
  • In various enzymatic assay using oxidase-peroxidase (such as Glucose oxidase system, Cholesterol oxidase system, Acyl coenzyme A oxidase system, and Glycerol-3-phosphate oxidase system), accuracy can be increased as the interference of bilirubin is removed by oxidation
  • It has also been used as an enzymatic catalyst for the cathode of biofuel cells that work under neutral conditions
TRUSTED_SUSTAINABILITY

Specifications

Color Light Bluish-Green
Content And Storage -20°C, Store Desiccated
Source Myrothecium Species
Quantity 20 U
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.