Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Bimetal Strip
SDPSCI_ED

Catalog No. S07008 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07008 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07008 Supplier United Scientific Supplies Supplier No. BMST05
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrate the thermal properties of matter.

  • Bimetallic bar bends when heated
  • Wooden handle
  • Activity sheet included

Specifications

Grade 6 to 12
For Use With (Application) To demonstrate the thermal properties of matter
Includes Activity sheet
Dimensions (L x W x H) 7.48 x 0.98 x 0.98 in. (19 x 2.5 x 2.5 cm)
Product Type Bimetallic Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.