Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Bimetallic Commercial Strips
SCI_ED

Catalog No. S23744 Shop All Eisco Products
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S23744 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23744 Supplier Eisco™ Supplier No. PH0382
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pack of Bimetallic Commercial Strips for practicals and project work.

  • Students investigate bending action by heating strips.
  • Strips can be built into projects such as thermostat, fire alarms, frost alarm, thermometers, etc.
  • Pack of 10 strips measuring 100 x 8 x 0.5 mm.
  • Strips securely attached to wooden supports.

Specifications

Grade 7 to 12
For Use With (Application) For practicals and project work
Includes Bi-metal strips
Quantity 10 Pk.
Dimensions (L x W x H) 100 x 8 x 0.5 mm
Product Type Bimetallic Commercial Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.