Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534687 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534687 Supplier Invitrogen™ Supplier No. PA534687
  • $652.00
    $508.65 / Each of 1

    Save $143.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: A431, mouse kidney. Predicted reactivity: Mouse (92%), Rat (80%), Pig (92%), Chicken (81%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes several isoforms of a nucleocytoplasmic adaptor protein, one of which was initially identified as a MYC-interacting protein with features of a tumor suppressor. Isoforms that are expressed in the central nervous system may be involved in synaptic vesicle endocytosis and may interact with dynamin, synaptojanin, endophilin, and clathrin. Isoforms that are expressed in muscle and ubiquitously expressed isoforms localize to the cytoplasm and nucleus and activate a caspase-independent apoptotic process. Studies in mouse suggest that this gene plays an important role in cardiac muscle development. Alternate splicing of the gene results in ten transcript variants encoding different isoforms. Aberrant splice variants expressed in tumor cell lines have also been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene BIN1
Gene Accession No. O00499, O08539
Gene Alias ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9
Gene Symbols BIN1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BIN1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 274, 30948
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.