Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Binding Buffer (BBA) for ReliaPrep™ Large Volume HT gDNA Isolation System

Catalog No. PRA1741 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1.6 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA1741 1.6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA1741 Supplier Promega Supplier No. A1741
Add to Cart
Add to Cart

Binding Buffer (BBA) is a component of the ReliaPrep Large Volume HT gDNA Isolation System (Cat.# A2751) and Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671) available as a standalone item.

  • Reagent Used in DNA Purification Protocols
  • Component of the ReliaPrep™ Large Volume HT gDNA Isolation System (Cat.# A2751) available as a standalone item
  • Component of the Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671) available as a standalone item

Specifications

Content And Storage 15°C to 30°C
Quantity 1.6 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.