Learn More
Essendant BINDINGPLAS1/4100/PKBK
Supplier:  Essendant GBC4000020
Encompass_Preferred
Communicate your ideas with professional polish 19-ring plastic spines won t scratch chip peel or discolor Bind 11x8 1/2 documents Editing capability with a machine Compatible with GBC and other plastic comb binding machines To select correct diameter measure thickness of material (including covers) and add 1/8 Value Pack Stock up and save Number of Rings- 19 Spine Capacity- 0 25 Sheet Capacity- 25 Document Size Range- Letter
Catalog No. 50-000-34205
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.