Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Bio-bin™ Waste Disposal Containers
SCI_ED

Catalog No. S29244 Shop All Dynalab Corporation Products
Change view
Click to view available options
Dimensions (L x W x H):
11 x 11 x 16.3 in.
16.85 x 7.12 x 11.25 in.
6 x 6 x 15 in.
6.7 x 6.7 x 7.87 in.
7.1 x 10.8 x 12.32 in.
9 x 4.3 x 4.7 in.
Capacity (Metric):
2.5 L
6 L
30 L
30L
Product Type:
Disposal Container
M Stand
U Stand
Z Stand
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Capacity (Metric) Product Type
S29244 11 x 11 x 16.3 in. 30 L Disposal Container
S29243 6 x 6 x 15 in. 6 L Disposal Container
S29246 6.7 x 6.7 x 7.87 in. 6 L Z Stand
S29242 9 x 4.3 x 4.7 in. 2.5 L Disposal Container
S29245 7.1 x 10.8 x 12.32 in. 6 L U Stand
S29247 16.85 x 7.12 x 11.25 in. 30L M Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S29244 Supplier Dynalon™ Supplier No. 7973030030/EMD
Only null left
Add to Cart
Edge
Add to Cart

Bio-bin™ is the ideal solution for biohazard or non-hazardous waste disposal. Reduce cost, weight, transport, CO2 emissions, storage space!

  • Space efficient, lightweight fold-flat design
  • Built-in lid and handles
  • Temporary and permanent seals
  • Stackable post use
  • UN 3291 Approved
  • Printed user instructions and user ID/date information on lid
  • Waterproof construction 

Specifications

Capacity (Metric) 30 L
Certifications/Compliance UN 3291
Closure Type Seal
Capacity (English) 7.925 gal.
Color White, Green, Orange
Dimensions (L x W x H) 11 x 11 x 16.3 in.
Height (Metric) 414.02 mm
Length (Metric) 279.4 mm
Shape Rectangle
Width (Metric) 279.4 mm
Autoclavable Autoclavable
Includes 2x2 in. Biohazard Waste Labels, 1.5mL thick PE bag, Absorbent Bio-Matt
Liner Material Polyethylene
For Use With (Application) Biohazard or non-hazardous waste
Width (English) 11 in.
Length (English) 11 in.
Height (English) 16.3 in.
Product Type Disposal Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.