Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio/Data™ Test Tubes
SDP

Catalog No. 22515278
Encompass
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
7.25 mm
Length (Metric):
55 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
22-515-278 7.25 mm 55 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-515-278 Supplier Bio/Data™ Supplier No. 101521
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Length (Metric) 55 mm
Material Siliconized Glass
For Use With PAP-4 series platelet aggregometers
Includes Silicone Coating
Diameter (Metric) 7.25 mm
Product Type Microvolume Test Tube
Bottom Shape Flat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.