Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig Bio-Fluids Solidifying Absorbent Powder
SDP

Catalog No. 18389141 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Co-Polymer Proprietary Blend
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-389-141 Co-Polymer Proprietary Blend
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-389-141 Supplier New Pig Supplier No. PLP503
  • $449.00
    $387.65 / Case of 4

    Save $61.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fast-acting absorbent captures biofluids like blood, urine and vomit and locks them inside for cleaner, safer disposal.

  • Permanently solidifies to lock in absorbed liquids so they won't leak or liquefy when the absorbents are handled or placed under pressure
  • Contains chlorine to kill pathogens, reduce odors and help you meet OSHA standards
  • Clean scent helps reduce offensive odors during and after cleanup Environmentally-friendly product absorbs biological fluids including blood, urine and vomit
  • Forms a solid, non-expanding mass for easier, less-expensive recovery and cleanup
  • May help you meet EPA requirements when solidifying waste before shipment or landfilling
  • Shaker bottle makes it easy to apply absorbent directly onto spill area
  • High Btu value produces less than 0.1% ash content, ideal for fuels blending and incineration

Specifications

Capacity (English) Sorbing Each absorbs up to 8 gal.
Color White
Material Co-Polymer Proprietary Blend
Type Absorbent Powder
For Use With (Application) Fast-acting absorbent captures biofluids like blood, urine and vomit and locks them inside for cleaner, safer disposal.
Includes 4 containers per box
Weight (English) 4 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.