Learn More
Fisherbrand™ Multi-Cap Tool
Catalog No. 054074 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
05-407-4 Polypropylene
1 options
This item is not returnable. View return policy
Catalog No. 05-407-4
Add to Cart
Add to Cart
This item is not returnable. View return policy
Prevents fatigue, saves wear on fingers and thumb.
  • Assists in the firm closing, sealing and opening of 0.2mL and 0.5mL thin wall microtubes
  • Prevents cross contamination and aerosolizing
  • Polypropylene
Specifications
Material Polypropylene
For Use With (Application) Closing and opening of micro reaction tubes