Learn More
Bio Plas™ Uni-Capsettes™ Process/Embedding Cassettes
Catalog No. 0270639
Change view
Click to view available options
Color:
Burnt Orange
Lilac
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-706-39 Yellow
02-706-41 Burnt Orange
02-706-38 White
02-706-42 Lilac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-706-39 Supplier Bio Plas™ Supplier No. 6051
Add to Cart
Add to Cart
Provides a modular and organized means for safe and consistent processing, embedding, sectioning, and storage of specimen samples
  • Detachable plastic lids
  • Heavy wall construction will not float or open during processing
  • Engineered to eliminate“chatter”during sectioning
  • Strategically designed slots allow for efficient solvent flow and drainage
  • Special acetyl copolymer plastic resists heat, solvents, decacifying solutions, and problems with microwave processing
  • Fit standard stainless steel and disposable plastic base molds
  • Accepted by all current automatic cassette labelers
Specifications
Color Yellow
Material Acetal Copolymer Plastic