Learn More
Bio Plas™ Uni-Flex™ Safety Caps
Description
Protects laboratory staff from infectious aerosols
- Liquid-tight seal ensures sample integrity and prevent spillage of serum samples
- Designed to hold under centrifugation, agitation, refrigeration and freezing
- Stretchable, unique, over-locking design to prevent evaporation
- One-hand application and removal
- Trace-metal free material
Specifications
Specifications
| Includes | Liquid-tight seal |
| Product Type | Safety Cap |
| Material | Polyethylene |
| Color | Red |
| For Use With (Equipment) | 16 mm Culture and Blood Collection Tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.