Learn More
Bio-Serv™ AIN-76 Rodent Liquid Animal Diet

Description
- Easy to prepare
- Offers the versatility of a liquid format, allowing the addition of alcohol, medications, or test compounds
- Provides a convenient way to measure food intake or study nutritional manipulation
- Ideal for post-operative recovery
- Nutritionally assayed
- Powder
- Ingredients: Corn Syrup Solids, Casein, Corn Oil, Cellulose, Maltodextrin, Mineral Mix, Suspending Aids, Vitamin Mix, DL-Methionine, Choline Bitartrate
Specifications
Specifications
| Product Type | Rodent Liquid Diet (Control Shake and Pour Bottle) |
| Components | Corn syrup solids, Casein, Corn Oil, Cellulose, Maltodextrin, Mineral Mix, Suspending aids, Vitamin Mix, DL-Methionine, Choline Bitartrate |
| Format | Powder |
| For Use With (Application) | Post operative recovery |
| Particle Shape | Powder |
| Quantity | 4 L |
| Species | Rodent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.