Learn More
Bio-Serv™ Bacon Softies™ Animal Diet, Certified

Description
- Soft, pelleted, nutritionally rich diet for rodents and supplement for canines
- Certified (contaminant screened)
- Nutritionally assayed
Ingredients: Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA
Specifications
Specifications
| Product Type | Animal Diet |
| Components | Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA |
| Flavor | Bacon |
| Format | Pellet |
| For Use With (Application) | For recovering or debilitated animals and weanlings |
| Particle Shape | Soft Pellet |
| Quantity | 1000 g |
| Species | Rodent, Canine |
| Sterility | Sterile |
| Diameter (English) | 0.5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.