Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bio-Serv™ Bacon Yummies™ Enrichment Treats, Certified

Nutritionally complete grain-based treat with a highly palatable bacon flavor
$53.05 - $155.78
Specifications
Product Type | Bacon Yummies |
---|---|
Calories | 3.37 |
Components | Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin |
Flavor | Bacon |
Format | Pellet |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-726-579
![]() ![]() |
Bio-Serv™
F057781 |
5000/Bg |
Each for $53.05
|
|
|||||
14-726-580
![]() ![]() |
Bio-Serv™
F05778 |
20,000/Bag |
Each for $155.78
|
|
|||||
Description
- Excellent for reward or foraging
- Can be fed as a sole source diet for animals with poor appetites
- 190mg size eliminates guesswork when measuring food intake
- Nutritionally assayed
- Certified (Contaminant Screened)
- Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
Specifications
Bacon Yummies | |
Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin | |
Pellet | |
Cool dry place |
3.37 | |
Bacon | |
Pellet |
Safety and Handling
ShelfLife : 6 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title